Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Anti-CD25/IL2RA Antibody (FITC), Rabbit Monoclonal 1 * 100 Tests
Catalog # SINO10165R216F-100
Supplier:
Sino Biological
Expand image
Recombinant Anti-CD25/IL2RA Antibody (FITC), Rabbit Monoclonal 1 * 100 Tests
Catalog # SINO10165R216F-100
Supplier:
Sino Biological
Frequently Bought Together
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Spare Lid of Ice Bucket 2.5 or 4.5 L, colour option, RED, BLUE, GREEN, BLACK 1 * 1 items
Supplier: VWR Collection
Cryoboxes, B01/B02/B03/B40/B60/B70/B90-series, without divider
Supplier: GLW
Strips of 8 PCR tubes with detached cap strips
Supplier: Brand
Screw vials free-standing sterile, gradu 1 * 500 items
Supplier: Roth Carl
Caps for roller bottles
Supplier: Corning
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Cryo Boxes
Supplier: VWR Collection
Corning® HYPERStack® Nest
Supplier: Corning