0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Recombinant Rhesus IL-18R beta / IL-18RAP Protein (His Tag), HPLC-verified 1 * 100 µG
SINO90122-C08H-100
:
Sino Biological
Recombinant Rhesus IL-18R beta / IL-18RAP Protein (His Tag), HPLC-verified 1 * 100 µG
SINO90122-C08H-100
:
Sino Biological
Frequently Bought Together
Spreader,L-shaped,sterile,plastic,(in inner pack of 5) 1 * 1.000 items
: TSC LIMITED
Transfer pipettes, general purpose, Samco™
: SAMCO
Corning® HYPERStack® Nest
: Corning
PCR plates, 96-well, fully skirted, low profile, Thermo-Fast®
: Thermo Fisher Scientific
Accessories for laboratory blenders, BagTools®
: INTERSCIENCE
Pierce™ Immunoassay Microplates, Steptavidin Coated, 96-Well
: Thermo Fisher Scientific
Inoculating loops and needles
: SARSTEDT
PCR tubes, 0,1 ml, with or without cap strips, Axygen®
: Corning
PCR plates for Roche® LightCycler® 480, Axygen®
: Corning