Human respiratory syncytial virus (RSV) Glycoprotein G Protein (aa 64-321, His Tag) 1 * 100 µG
Catalog # SINO40626-V08H-100
Supplier: Sino Biological
Human respiratory syncytial virus (RSV) Glycoprotein G Protein (aa 64-321, His Tag) 1 * 100 µG
Catalog # SINO40626-V08H-100
Supplier: Sino Biological
Frequently Bought Together
MicroWell™ Plates, 96 well, Nunc-Immuno™
Supplier: Thermo Fisher Scientific
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Cell culture plates, 96-well, Corning®
Supplier: Corning
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
Scoops
Supplier: VITLAB
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Supplier: Corning
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
96 MicroWell™ plates, Nunclon™∆
Supplier: Thermo Fisher Scientific
Screw caps for laboratory/media bottles
Supplier: SciLabware