0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Recombinant Human Siglec-3/CD33 Protein (His Tag), Amine-Labeled Biotinylated 1 * 100 µG
SINO12238H08HB-100
:
Sino Biological
Recombinant Human Siglec-3/CD33 Protein (His Tag), Amine-Labeled Biotinylated 1 * 100 µG
SINO12238H08HB-100
:
Sino Biological
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR® Blender Bags
: VWR Collection
Transfer pipettes
: RATIOLAB
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection