Recombinant Human Vitamin D Receptor/VDR Protein (His Tag) 1 * 50 µG
Catalog # SINO12025-H08B-50
Supplier: Sino Biological
Recombinant Human Vitamin D Receptor/VDR Protein (His Tag) 1 * 50 µG
Catalog # SINO12025-H08B-50
Supplier: Sino Biological
Frequently Bought Together
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
VWR®, Cell Lifters
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Containers, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Petri dishes
Supplier: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Cryoboxes
Supplier: RATIOLAB
VWR® Cell Culture Dishes
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Sampling scoops, PS, white, double-bagged, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
PARTITION INSERTS FOR CRYOBOX 10X10 1 * 10 items
Supplier: Roth Carl