0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Recombinant Human Apolipoprotein H Protein (His Tag), HPLC-verified 1 * 100 µG
SINO11221-H08H-100
:
Sino Biological
Recombinant Human Apolipoprotein H Protein (His Tag), HPLC-verified 1 * 100 µG
SINO11221-H08H-100
:
Sino Biological
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Multiwell Cell Culture Plates
: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Petri dishes
: Anicrin
VWR® Cellulose Stoppers
: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
: GLW
Petri dishes
: Greiner Bio-One
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Flat Screw Caps for Microcentrifuge Tubes
: VWR Collection
Cap strips, flat top
: Thermo Fisher Scientific
Screw caps for micro tubes
: SARSTEDT
Cryoboxes, coloured, Nalgene®
: Thermo Fisher Scientific