Recombinant Human IRAK4 Protein (His Tag) 1 * 500 µG
Catalog # SINO10735-H07B-500
Supplier: Sino Biological
Recombinant Human IRAK4 Protein (His Tag) 1 * 500 µG
Catalog # SINO10735-H07B-500
Supplier: Sino Biological
Frequently Bought Together
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Blotting Pads, Grade 707
Supplier: VWR Collection
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Cryobox B68 1 * 1 items
Supplier: GLW
FastPrep® Empty Tubes and Caps
Supplier: MP Biomedicals
AggreWellTrademark400 24-well plate, 1 pack 1 * 1 items
Supplier: STEMCELL Technologies
Cell factories, Nunc™ high density
Supplier: Thermo Fisher Scientific
PCR plates, 96-well
Supplier: Brand