Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Human IRAK4 Protein (His Tag) 1 * 50 µG
Catalog # SINO10735-H07B-50
Supplier:
Sino Biological
Expand image
Recombinant Human IRAK4 Protein (His Tag) 1 * 50 µG
Catalog # SINO10735-H07B-50
Supplier:
Sino Biological
Frequently Bought Together
VWR®, Test Tubes without Rim
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Transfer pipettes
Supplier: RATIOLAB
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection