Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Human EpCAM/TROP1 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified 1 * 20 µG
Catalog # SINO10694H08H-B-20
Supplier:
Sino Biological
Expand image
Recombinant Human EpCAM/TROP1 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified 1 * 20 µG
Catalog # SINO10694H08H-B-20
Supplier:
Sino Biological
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Transfer pipettes
Supplier: RATIOLAB
Serological pipettes, Stripette™
Supplier: Corning
VWR® Blender Bags
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF