Recombinant Human IL-18R beta / IL-18RAP Protein (His Tag), HPLC-verified 1 * 100 µG
SINO10176-H08H-100
Recombinant Human IL-18R beta / IL-18RAP Protein (His Tag), HPLC-verified 1 * 100 µG
SINO10176-H08H-100
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
Transfer pipettes
: RATIOLAB
VWR® Standard Line Transfer Pipettes
: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Blender Bags
: VWR Collection
VWR®, Straw Pipettes
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection