Recombinant Human Complement component 3 Protein (His Tag), HPLC-verified 1 * 1 mg
Catalog # SINO13182-H08H-1
Supplier: Sino Biological
Recombinant Human Complement component 3 Protein (His Tag), HPLC-verified 1 * 1 mg
Catalog # SINO13182-H08H-1
Supplier: Sino Biological
Frequently Bought Together
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
Sterilin™ Containers, Bijou, with Screw Cap
Supplier: STERILIN
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® Bottles, Water Sampling
Supplier: VWR Collection
Multiple well cell culture plates, Costar®
Supplier: Corning
Cell culture flasks with standard or filter caps, CELLSTAR®
Supplier: Greiner Bio-One
VWR® Cell Culture Dishes
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Internal thread cryogenic vials
Supplier: Corning
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Sample tubes
Supplier: Brand
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One