Recombinant Mouse CD98 Protein (His Tag), HPLC-verified 1 * 50 µG
Catalog # SINO50813-M07H-50
Supplier: Sino Biological
Recombinant Mouse CD98 Protein (His Tag), HPLC-verified 1 * 50 µG
Catalog # SINO50813-M07H-50
Supplier: Sino Biological
Frequently Bought Together
VWR®, Drosophila Stock Bottles
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Deep well plates and cover mats
Supplier: Brand
VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
Accessories for Q Cyclers
Supplier: Quantabio
Eppendorf LoBind® twin.tec® PCR Plates
Supplier: EPPENDORF
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
Matrix™ Tri-Coded Tubes
Supplier: THERMO MATRIX TECHNOLOGIES
Transfer pipettes, fine tip
Supplier: Alpha Laboratories
Sampling spoons curved, long handle, LaboPlast®/SteriPlast®
Supplier: BURKLE
Microplates, NeutrAvidin™ coated, 96- and 384- well, Pierce™
Supplier: Thermo Fisher Scientific
Strips of 8 PCR tubes and cap strips
Supplier: RATIOLAB
PCR tubes, with flat or domed cap, Sapphire
Supplier: Greiner Bio-One
384-well (HTS) microplates, non treated, BRANDplates®
Supplier: Brand
Microplates, 384-well
Supplier: Greiner Bio-One