Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Human GITR/TNFRSF18/AITR Protein (ECD, His Tag), HPLC-verified 1 * 1 mg
Catalog # SINO13643-H08H-1
Supplier:
Sino Biological
Expand image
Recombinant Human GITR/TNFRSF18/AITR Protein (ECD, His Tag), HPLC-verified 1 * 1 mg
Catalog # SINO13643-H08H-1
Supplier:
Sino Biological
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Test Tubes without Rim
Supplier: VWR Collection
Petri dishes
Supplier: Greiner Bio-One
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection