Recombinant Human CD63 Protein (ECD, His Tag) 1 * 1 mg
Catalog # SINO11271-H08H-1
Supplier: Sino Biological
Recombinant Human CD63 Protein (ECD, His Tag) 1 * 1 mg
Catalog # SINO11271-H08H-1
Supplier: Sino Biological
Frequently Bought Together
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
Lids for Nunc™ microplates
Supplier: Thermo Fisher Scientific
Cell culture plates, Nunc™ Multidish
Supplier: Thermo Fisher Scientific
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
Containers, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cell culture flasks, Corning®
Supplier: Corning
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
MicroWell™ Plates, 96 well, Nunc-Immuno™
Supplier: Thermo Fisher Scientific
Sample tubes for stool, with screw cap
Supplier: BOETTGER PAUL
ViewPlate-384 blk,clr btm,Tissue Culture 1 * 40 items
Supplier: PerkinElmer
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Supplier: EPPENDORF
Matrix™ Reagent Reservoirs
Supplier: THERMO MATRIX TECHNOLOGIES