SODIUM BIFLUORIDE TECNICAL 1 * 2,5 kg
Catalog # SPCMS1160-2.5KG
Supplier: Spectrum Chemical
Frequently Bought Together
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
Disodium hydrogen phosphate dodecahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium formate ≥97%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals