Order Entry
Ireland
ContactUsLinkComponent
Straight threaded coupling, PFA, 1/2" NPT(F)
Straight threaded coupling, PFA, 1/2" NPT(F)
Catalog # MFLX06546-03
undefined
Straight threaded coupling, PFA, 1/2" NPT(F)
Catalog # MFLX06546-03
About this item
Create a secure pipe threaded connection.
  • Constructed of chemically inert PFA
These straight threaded coupling fittings are made of chemically inert PFA. Fittings can be sterilized via autoclave or ethylene oxide (ETO).
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 1):
    ½"
  • Thread size NPT(F) (port 2):
    ½"
  • Description:
    Straight threaded coupling, PFA, ½" NPT(F)
  • Environmentally Preferable:
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Warranty:
    1 year
  • Pk:
    1
  • Fitting material:
    PFA
Frequently Bought Together