Order Entry
Ireland
ContactUsLinkComponent
Masterflex® Flow-Through Tubing Weight; Stainless Steel
Masterflex® Flow-Through Tubing Weight; Stainless Steel
Catalog # MFLX07570-04
undefined
Masterflex® Flow-Through Tubing Weight; Stainless Steel
Catalog # MFLX07570-04
About this item
Keep your samples flowing and tubing from curling.
  • Choose between PTFE or stainless steel construction
This flow-through tubing weight prevents tubing from curling or floating. Ideal for groundwater sampling, this weight fits Masterflex®; L/S® 15 or L/S® 24 Precision tubing.
  • Description:
    Stainless steel flow-through tubing weight. 1 year warranty
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Pk:
    1
  • Max. int.Ø:
    0,25"
  • Min. int.Ø:
    6,8 mm (0,1875")
Frequently Bought Together