FIX RAL 555 1 * 1 L
Catalog # 720-0361
Supplier: RAL
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals