ERK1/2 (Total) and ERK1/2 (Phospho) TR-FRET Assay Kit 1 * 1 KIT
CAYM500208-480
Frequently Bought Together
Transfer pipettes
: RATIOLAB
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Ethanol, GPR RECTAPUR®
: VWR Chemicals
VWR®, Petri Dish, PS
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection