SARS-CoV/SARS-CoV-2 Nucleocapsid Protein Rabbit Monoclonal Antibody (Clone 0002) 1 * 100 µl
Catalog # CAYM33628-100
Supplier: Cayman Chemical
SARS-CoV/SARS-CoV-2 Nucleocapsid Protein Rabbit Monoclonal Antibody (Clone 0002) 1 * 100 µl
Catalog # CAYM33628-100
Supplier: Cayman Chemical
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Ethylene glycol 99.5% for analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific