MMP-12 (C-Term) Rabbit Monoclonal Antibody 1 * 100 µl
CAYM32305-100
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals