4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazole 1 * 25 mg
Catalog # PROOMM0939.01-0025
Supplier: LGC Standards PROMOCHEM
4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazole 1 * 25 mg
Catalog # PROOMM0939.01-0025
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Benzoic acid, Supelco®
Supplier: Merck
Redox buffer solutions
Supplier: Mettler - Toledo
pH buffer, single-use, SINGLET™
Supplier: Hach
pH Standard Solutions, IUPAC, Certified
Supplier: Hach