Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 Spike NTD His Tag (B.1.1.529/Omicron) (SPD-C522d) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Le u 303 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE)). The spike mu entifi 1 * 100 µG
Catalog # ACRBSPDC522D-100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike NTD His Tag (B.1.1.529/Omicron) (SPD-C522d) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Le u 303 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE)). The spike mu entifi 1 * 100 µG
Catalog # ACRBSPDC522D-100UG
Supplier:  ACRO Biosystems
Frequently Bought Together