PLP381multi behrotest peristaltic pump capacity 0,0025...380 ml/min without pump head 1 * 1 items
Catalog # BEHRB00738276
Supplier: BEHR
PLP381multi behrotest peristaltic pump capacity 0,0025...380 ml/min without pump head 1 * 1 items
Catalog # BEHRB00738276
Supplier: BEHR
Frequently Bought Together
VWR®, Single-Use Heads for Air Samplers
Supplier: VWR Collection
VWR® pHenomenal® pH 1100L/1100LB pH/mV/°C Meter, Bench
Supplier: VWR Collection
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Multi-parameter meters, handheld, 3320
Supplier: WTW
VWR® HCO 304, Conductivity Meter, Handheld
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR®, Square Cuvettes
Supplier: VWR Collection
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
VWR®, Nitrile Gloves
Supplier: VWR Collection
Accessories for cell counter, EVE™
Supplier: NANOENTEK
Titration beakers
Supplier: Mettler - Toledo
Accessories for titrators
Supplier: Mettler - Toledo
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Plastic cuvette for LICO
Supplier: Hach
Cell culture reagents, microcarriers, Cytodex 3
Supplier: Cytiva