Order Entry
Ireland
ContactUsLinkComponent
3/4 HYGIENIC CLAMP SEAL (EDPM) ELAST-O-PURE EP75 BLACK USP 34 CLASS VI.FDA 21 CFR 177.2600 CONFORMING (AQUEOUS ONLY) WITHOUT OUTER LIP NS (IN). 75 DRAWING NO. JW 197166 BAGGED IN 25's 1 * 25 items
Catalog # IBSAS000087611
Supplier:  IBS Stock Items
3/4 HYGIENIC CLAMP SEAL (EDPM) ELAST-O-PURE EP75 BLACK USP 34 CLASS VI.FDA 21 CFR 177.2600 CONFORMING (AQUEOUS ONLY) WITHOUT OUTER LIP NS (IN). 75 DRAWING NO. JW 197166 BAGGED IN 25's 1 * 25 items
Catalog # IBSAS000087611
Supplier:  IBS Stock Items
Frequently Bought Together