Order Entry
Ireland
ContactUsLinkComponent
Nicolet iN5 IR Microscope with DTGS Including: 10x IR objective 0.71 N.A. 10x IR condenser Manual x,y stage Manual course and fine focusing LED illumination Transmission, Reflection modes Basic sampling tool kit Fixed 100 micron apert 1 * 1 items
Catalog # THEC912A0922
Supplier:  THERMO EC
Nicolet iN5 IR Microscope with DTGS Including: 10x IR objective 0.71 N.A. 10x IR condenser Manual x,y stage Manual course and fine focusing LED illumination Transmission, Reflection modes Basic sampling tool kit Fixed 100 micron apert 1 * 1 items
Catalog # THEC912A0922
Supplier:  THERMO EC
Frequently Bought Together