Order Entry
Ireland
Orders LinkContactUsLinkComponent
Add-on storage units
Add-on storage units
Table add-on 1 drawer with lock, 452×516×152 mm
Catalog # TEKMTCA0100
Supplier:   TEKNOMEK
Add-on storage units
Catalog # TEKMTCA0100
Supplier:  TEKNOMEK
Add-on storage units
Add-on storage units
Table add-on 1 drawer with lock, 452×516×152 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
304 grade stainless steel add on unit for rear railed workbench is perfect for adding additional storage. Cannot be sold singly.
  • Fully welded 304-grade stainless steel
  • Brushed dull polish finish
Supplied fitted to workbench when ordered together. Choice of one drawer, two drawers, three drawers or cupboard unit, for our rear railed workbenches. Enables the user to create a workstation to suit their individual requirements.
Specifications
  • Description:
    Table add-on 1 drawer with lock
  • Seq:
    03
  • W×D×H:
    452×516×152 mm