Order Entry
Ireland
ContactUsLinkComponent
AR Diamond Crystal Plate for Smart iTX/iD7/Everest ATR Single Reflection Diamond Crystal Plate - High Throughput Spectral range with KBr/DTGS optics: 7,800 - 400 cm-1 Best overall performance monolithic diamond crystal Includes a 5-year parts 1 * 1 items
Catalog # THEC869-168800
Supplier:  THERMO EC
AR Diamond Crystal Plate for Smart iTX/iD7/Everest ATR Single Reflection Diamond Crystal Plate - High Throughput Spectral range with KBr/DTGS optics: 7,800 - 400 cm-1 Best overall performance monolithic diamond crystal Includes a 5-year parts 1 * 1 items
Catalog # THEC869-168800
Supplier:  THERMO EC
Frequently Bought Together