Order Entry
Ireland
ContactUsLinkComponent
Calibration of test sieves with woven metal wire cloth. Calibration according to DIN ISO 3310-1. By accredited calibration laboratory according to ISO/IEC 17025:2005. Calibration mark D-K-18542-01-00 for nominal diameters D1 up to 203 MM 1 * 1 items
Catalog # HAVE206697271
Supplier:  NEXOPART
Calibration of test sieves with woven metal wire cloth. Calibration according to DIN ISO 3310-1. By accredited calibration laboratory according to ISO/IEC 17025:2005. Calibration mark D-K-18542-01-00 for nominal diameters D1 up to 203 MM 1 * 1 items
Catalog # HAVE206697271
Supplier:  NEXOPART
Frequently Bought Together