Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human CD3 epsilon, His,Avitag (CDE-H82E1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 ( Accession # P07766-1). 1 * 200 µG
Catalog # ACRBCDE-H82E1200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD3 epsilon, His,Avitag (CDE-H82E1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 ( Accession # P07766-1). 1 * 200 µG
Catalog # ACRBCDE-H82E1200UG
Supplier:  ACRO Biosystems
Frequently Bought Together