Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein (R203K, G204R), His Tag (NUN-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (R203K, G204R)). The nucleocapsid protein is consisted of the N-terminal RNA-bindi 1 * 100 µG
Catalog # ACRBNUN-C52HG100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein (R203K, G204R), His Tag (NUN-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (R203K, G204R)). The nucleocapsid protein is consisted of the N-terminal RNA-bindi 1 * 100 µG
Catalog # ACRBNUN-C52HG100UG
Supplier:  ACRO Biosystems
Frequently Bought Together