Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid Protein NTD, His Tag (NUN-C5143) is expressed from E.coli cells. It contains AA Gly 44 - Glu 174 (Accession # QHO62115.1). 1 * 1 mg
Catalog # ACRBNUN-C5143-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid Protein NTD, His Tag (NUN-C5143) is expressed from E.coli cells. It contains AA Gly 44 - Glu 174 (Accession # QHO62115.1). 1 * 1 mg
Catalog # ACRBNUN-C5143-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together