Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (NUN-C5145) is expressed from E.coli cells. It contains AA Ser 255 - Pro 364 (Accession # QHO62115.1). 1 * 100 µG
Catalog # ACRBNUN-C5145100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (NUN-C5145) is expressed from E.coli cells. It contains AA Ser 255 - Pro 364 (Accession # QHO62115.1). 1 * 100 µG
Catalog # ACRBNUN-C5145100UG
Supplier:  ACRO Biosystems
Frequently Bought Together