Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 Spike NTD (T95I, G142D, E154K), His Tag (S1D-C52Hf) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - L eu 303 (Accession # QHD43416.1 (T95I, G142D, E154K)). The mutations (T95I, G142D, E154K) were identified in the SARS- 1 * 1 mg
Catalog # ACRBS1D-C52HF-1MG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike NTD (T95I, G142D, E154K), His Tag (S1D-C52Hf) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - L eu 303 (Accession # QHD43416.1 (T95I, G142D, E154K)). The mutations (T95I, G142D, E154K) were identified in the SARS- 1 * 1 mg
Catalog # ACRBS1D-C52HF-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together