Order Entry
Ireland
ContactUsLinkComponent
The Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Spike RBD F(ab')2. 1 * 200 µG
Catalog # ACRBSPD-BM227200UG
Supplier:  ACRO Biosystems
The Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Spike RBD F(ab')2. 1 * 200 µG
Catalog # ACRBSPD-BM227200UG
Supplier:  ACRO Biosystems
Frequently Bought Together