Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human Siglec-2, His,Avitag (SI2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Acc ession # P20273-1). 1 * 200 µG
Catalog # ACRBSI2-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human Siglec-2, His,Avitag (SI2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Acc ession # P20273-1). 1 * 200 µG
Catalog # ACRBSI2-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together