Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human LIV-1, His,Avitag (LV1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Access ion # Q13433-1). 1 * 25 µG
Catalog # ACRBLV1-H82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human LIV-1, His,Avitag (LV1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Access ion # Q13433-1). 1 * 25 µG
Catalog # ACRBLV1-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together