Order Entry
Ireland
ContactUsLinkComponent
Mouse MCAM, His Tag (MCM-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Val 563 (Accession # Q8R2Y2-1). 1 * 1 mg
Catalog # ACRBMCM-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Mouse MCAM, His Tag (MCM-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Val 563 (Accession # Q8R2Y2-1). 1 * 1 mg
Catalog # ACRBMCM-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together