Order Entry
Ireland
ContactUsLinkComponent
Mouse Carbonic Anhydrase IX / CA9 (32-390), His Tag (CA9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Asp 390 (Accession # Q8VHB5-1). 1 * 1 mg
Catalog # ACRBCA9-M52H3-1MG
Supplier:  ACRO Biosystems
Mouse Carbonic Anhydrase IX / CA9 (32-390), His Tag (CA9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Asp 390 (Accession # Q8VHB5-1). 1 * 1 mg
Catalog # ACRBCA9-M52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together