Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Mouse CD40, Fc,Avitag (CD0-M82F3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accessio n # P27512-1). 1 * 200 µG
Catalog # ACRBCD0-M82F3200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse CD40, Fc,Avitag (CD0-M82F3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accessio n # P27512-1). 1 * 200 µG
Catalog # ACRBCD0-M82F3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together