Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus NKG2A, His Tag (NKA-C5245) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # Q68VD 2-1). 1 * 100 µG
Catalog # ACRBNKA-C5245100UG
Supplier:  ACRO Biosystems
Cynomolgus NKG2A, His Tag (NKA-C5245) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # Q68VD 2-1). 1 * 100 µG
Catalog # ACRBNKA-C5245100UG
Supplier:  ACRO Biosystems
Frequently Bought Together