Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus NKG2A, His Tag (NKA-C5245) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # Q68VD 2-1). 1 * 1 mg
Catalog # ACRBNKA-C5245-1MG
Supplier:  ACROBIOSYSTEMS
Cynomolgus NKG2A, His Tag (NKA-C5245) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # Q68VD 2-1). 1 * 1 mg
Catalog # ACRBNKA-C5245-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together