Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1). 1 * 100 µG
ACRBCD9-M52H3100UG
:
ACRO Biosystems
Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1). 1 * 100 µG
ACRBCD9-M52H3100UG
:
ACRO Biosystems
Frequently Bought Together
Serological pipettes, Stripette™
: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
MicroWell™ Plates, 96 well, Nunc-Immuno™
: Thermo Fisher Scientific
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection