Order Entry
Ireland
ContactUsLinkComponent
Human FGF19, Fc Tag (FG9-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Lys 216 (Accession # O95750-1). 1 * 1 mg
Catalog # ACRBFG9-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Human FGF19, Fc Tag (FG9-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Lys 216 (Accession # O95750-1). 1 * 1 mg
Catalog # ACRBFG9-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together