Order Entry
Ireland
ContactUsLinkComponent
FITC-Labeled Human uPAR, His Tag (UPR-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1). 1 * 25 µG
Catalog # ACRBUPR-HF2H3-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human uPAR, His Tag (UPR-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1). 1 * 25 µG
Catalog # ACRBUPR-HF2H3-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together