Order Entry
Ireland
ContactUsLinkComponent
Human GPA33, Fc Tag (GP3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # Q99795-1). 1 * 1 mg
Catalog # ACRBGP3-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Human GPA33, Fc Tag (GP3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # Q99795-1). 1 * 1 mg
Catalog # ACRBGP3-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together