Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus / Rhesus macaque TIGIT, Mouse IgG2a Fc Tag (TIT-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # G7NXM4-1). 1 * 50 µG
Catalog # ACRBTIT-C5253-50UG
Supplier:  ACRO Biosystems
Cynomolgus / Rhesus macaque TIGIT, Mouse IgG2a Fc Tag (TIT-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # G7NXM4-1). 1 * 50 µG
Catalog # ACRBTIT-C5253-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together