Order Entry
Ireland
ContactUsLinkComponent
FITC-Labeled Human DLL3, His Tag (DL3-HF2H4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1). 1 * 25 µG
Catalog # ACRBDL3-HF2H4-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human DLL3, His Tag (DL3-HF2H4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1). 1 * 25 µG
Catalog # ACRBDL3-HF2H4-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together