Order Entry
Ireland
ContactUsLinkComponent
Canine PD-1, His Tag (PD1-C52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gly 168 (Accession # NP_001301 026.1). 1 * 100 µG
Catalog # ACRBPD1-C52H9100UG
Supplier:  ACRO Biosystems
Canine PD-1, His Tag (PD1-C52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gly 168 (Accession # NP_001301 026.1). 1 * 100 µG
Catalog # ACRBPD1-C52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together