Order Entry
Ireland
ContactUsLinkComponent
Mouse Angiopoietin-like 3, His Tag (AN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Thr 455 (Accessio n # Q9R182-1). 1 * 1 mg
Catalog # ACRBAN3-M52H3-1MG
Supplier:  ACRO Biosystems
Mouse Angiopoietin-like 3, His Tag (AN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Thr 455 (Accessio n # Q9R182-1). 1 * 1 mg
Catalog # ACRBAN3-M52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together